siRNA Details
| Gene Symbol: Prrc2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212462.2 | NP_997627.1 | 15 | 4036 | ||
| XM_006256048.2 | XP_006256110.1 | 16 | 4281 | |||
| XM_006256050.2 | XP_006256112.1 | 16 | 4281 | |||
| XM_006256051.2 | XP_006256113.1 | 16 | 4280 | |||
| XM_006256052.2 | XP_006256114.1 | 16 | 4281 | |||
| XM_006256053.2 | XP_006256115.1 | 16 | 4281 | |||

Prrc2a
Genomic Map