siRNA Details
| Gene Symbol: Ncr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181822.2 | NP_861543.2 | 2, 3 | 570 | ||
| XM_006256054.2 | XP_006256116.1 | 2, 3 | 740 | |||
| GenBank mRNA | AY273824.1 | - | 2, 3 | 691 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ncr3| Gene Symbol: Ncr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181822.2 | NP_861543.2 | 2, 3 | 570 | ||
| XM_006256054.2 | XP_006256116.1 | 2, 3 | 740 | |||
| GenBank mRNA | AY273824.1 | - | 2, 3 | 691 | ||