siRNA Details
Gene Symbol: Ncr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181822.2 | NP_861543.2 | 2 | 348 | ||
XM_006256054.2 | XP_006256116.1 | 2 | 518 | |||
GenBank mRNA | AY273824.1 | - | 2 | 469 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ncr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181822.2 | NP_861543.2 | 2 | 348 | ||
XM_006256054.2 | XP_006256116.1 | 2 | 518 | |||
GenBank mRNA | AY273824.1 | - | 2 | 469 |