siRNA Details
| Gene Symbol: Psmb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105727.2 | NP_001099197.2 | 2, 3 | 525 | ||
| GenBank mRNA | AI602737.1 | - | 1, 2 | 338 | ||
| D45247.1 | - | 2, 3 | 523 | |||
| FQ214283.1 | - | 1, 2 | 59 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psmb5| Gene Symbol: Psmb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105727.2 | NP_001099197.2 | 2, 3 | 525 | ||
| GenBank mRNA | AI602737.1 | - | 1, 2 | 338 | ||
| D45247.1 | - | 2, 3 | 523 | |||
| FQ214283.1 | - | 1, 2 | 59 | |||