siRNA Details
| Gene Symbol: Skiv2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213559.2 | NP_998724.1 | 24 | 2973 | ||
| XM_006255923.1 | XP_006255985.1 | 24 | 3036 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Skiv2l| Gene Symbol: Skiv2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213559.2 | NP_998724.1 | 24 | 2973 | ||
| XM_006255923.1 | XP_006255985.1 | 24 | 3036 | |||