siRNA Details
| Gene Symbol: Rps4x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007600.1 | NP_001007601.1 | 1 | 481 | ||
| GenBank mRNA | BC086560.1 | - | 1 | 481 | 7303578 | |
| FQ220060.1 | - | 1 | 503 | |||
| FQ226324.1 | - | 1 | 504 | |||
| FQ230280.1 | - | 1 | 504 | |||
| X14210.1 | - | 1 | 502 | |||

LOC100362640
Genomic Map