siRNA Details
| Gene Symbol: Slc39a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008885.2 | NP_001008885.1 | 4 | 1018 | ||
| NM_001164744.1 | NP_001158216.1 | 5 | 759 | |||
| GenBank mRNA | BC079141.1 | - | 4 | 1018 | 7127902 | |
| FQ212335.1 | - | 5 | 726 | |||
| FQ212833.1 | - | 4 | 1046 | |||

Slc39a7
Genomic Map