siRNA Details
| Gene Symbol: Armc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113777.1 | NP_001107249.1 | 6 | 1004 | ||
| GenBank mRNA | BC158769.1 | - | 6 | 1004 | 7115619 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Armc12| Gene Symbol: Armc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113777.1 | NP_001107249.1 | 6 | 1004 | ||
| GenBank mRNA | BC158769.1 | - | 6 | 1004 | 7115619 | |