siRNA Details
| Gene Symbol: Pknox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013074.1 | NP_001013092.1 | 6, 7 | 784 | ||
| XM_006256187.2 | XP_006256249.1 | 7, 8 | 1819 | |||
| XM_006256188.2 | XP_006256250.1 | 8, 9 | 1881 | |||
| XM_006256189.2 | XP_006256251.1 | 8, 9 | 1873 | |||
| XM_006256190.2 | XP_006256252.1 | 7, 8 | 1817 | |||
| XM_008772808.1 | XP_008771030.1 | 8, 9 | 1696 | |||
| XM_008772809.1 | XP_008771031.1 | 7, 8 | 1811 | |||
| XM_008772810.1 | XP_008771032.1 | 7, 8 | 1699 | |||
| GenBank mRNA | BC091397.1 | - | 6, 7 | 784 | 7323315 | |

Pknox1
Genomic Map