siRNA Details
| Gene Symbol: Trpm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011559.1 | NP_001011559.1 | 32 | 4423 | ||
| XM_006256248.2 | XP_006256310.1 | 33 | 4613 | |||
| XM_006256249.2 | XP_006256311.1 | 33 | 4610 | |||
| GenBank mRNA | AB193179.1 | - | 32 | 4451 | ||
| AY749166.1 | - | 32 | 4423 | |||

Trpm2
Genomic Map