siRNA Details
| Gene Symbol: Lrrtm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106387.1 | NP_001099857.1 | 2 | 1446 | ||
| XM_008772878.1 | XP_008771100.1 | 2 | 3122 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrtm3| Gene Symbol: Lrrtm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106387.1 | NP_001099857.1 | 2 | 1446 | ||
| XM_008772878.1 | XP_008771100.1 | 2 | 3122 | |||