siRNA Details
Gene Symbol: Por | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031576.1 | NP_113764.1 | 94 | |||
GenBank mRNA | M10068.1 | - | 1 | 58 | ||
M12516.1 | - | 94 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Por | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031576.1 | NP_113764.1 | 94 | |||
GenBank mRNA | M10068.1 | - | 1 | 58 | ||
M12516.1 | - | 94 |