siRNA Details
| Gene Symbol: Serinc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182951.1 | NP_891996.1 | 9 | 1068 | ||
| GenBank mRNA | AY339894.1 | - | 9 | 1068 | ||
| AY539942.1 | - | 4716 | ||||
| BC088852.1 | - | 9 | 1079 | 7315523 | ||
| DQ103708.1 | - | 9 | 1048 | |||

Serinc1
Genomic Map