siRNA Details
| Gene Symbol: Smpdl3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005539.1 | NP_001005539.1 | 8 | 1163 | ||
| GenBank mRNA | BC082029.1 | - | 8 | 1163 | 7130096 | |
| FQ230988.1 | - | 3 | 336 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smpdl3a| Gene Symbol: Smpdl3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005539.1 | NP_001005539.1 | 8 | 1163 | ||
| GenBank mRNA | BC082029.1 | - | 8 | 1163 | 7130096 | |
| FQ230988.1 | - | 3 | 336 | |||