siRNA Details
| Gene Symbol: Ddo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109465.2 | NP_001102935.1 | 3 | 255 | ||
| GenBank mRNA | AB983592.1 | - | 2 | 195 | ||
| Gene Symbol: LOC100911156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751978.3 | XP_003752026.1 | 3 | 317 | ||

Ddo
Genomic Map