siRNA Details
| Gene Symbol: Cox4i1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017202.1 | NP_058898.1 | 4, 5 | 453 | ||
| GenBank mRNA | BC084719.1 | - | 3, 4 | 369 | 7317602 | |
| FQ211185.1 | - | 4, 5 | 406 | |||
| FQ214265.1 | - | 4, 5 | 462 | |||
| FQ217878.1 | - | 4, 5 | 421 | |||
| FQ223981.1 | - | 4, 5 | 415 | |||
| FQ227128.1 | - | 4, 5 | 451 | |||
| FQ228805.1 | - | 4, 5 | 403 | |||
| X14209.1 | - | 4, 5 | 435 | |||
| X15029.1 | - | 3, 4 | 380 | |||

Cox4i1
Genomic Map