siRNA Details
| Gene Symbol: Ppa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100834.1 | NP_001094304.1 | 7 | 695 | ||
| GenBank mRNA | BC099794.1 | - | 7 | 695 | ||
| FQ215196.1 | - | 7 | 647 | |||
| FQ215675.1 | - | 7 | 647 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppa1| Gene Symbol: Ppa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100834.1 | NP_001094304.1 | 7 | 695 | ||
| GenBank mRNA | BC099794.1 | - | 7 | 695 | ||
| FQ215196.1 | - | 7 | 647 | |||
| FQ215675.1 | - | 7 | 647 | |||