siRNA Details
| Gene Symbol: Ascc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007632.2 | NP_001007633.1 | 8 | 840 | ||
| XM_006256404.2 | XP_006256466.1 | 8 | 1034 | |||
| XM_006256405.2 | XP_006256467.1 | 8 | 1031 | |||
| XM_006256407.2 | XP_006256469.2 | 8 | 920 | |||
| XM_006256408.2 | XP_006256470.1 | 8 | 989 | |||
| XM_006256409.1 | XP_006256471.1 | 7 | 775 | |||
| XM_006256410.2 | XP_006256472.1 | 8 | 986 | |||
| XM_008772881.1 | XP_008771103.1 | 7 | 772 | |||
| GenBank mRNA | BP504504.1 | - | 4 | 331 | ||
| FQ226270.1 | - | 5 | 487 | |||

Ascc1
Genomic Map