siRNA Details
| Gene Symbol: Wasf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025114.1 | NP_001020285.1 | 7, 8 | 974 | ||
| XM_006256586.2 | XP_006256648.1 | 7, 8 | 1167 | |||
| XM_006256587.2 | XP_006256649.1 | 7, 8 | 1165 | |||
| GenBank mRNA | BC091322.1 | - | 7, 8 | 974 | 7313558 | |

Wasf1
Genomic Map