siRNA Details
| Gene Symbol: Sgtb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181629.3 | NP_853660.1 | 6 | 535 | ||
| XM_006231882.2 | XP_006231944.1 | 6 | 701 | |||
| XM_006231883.2 | XP_006231945.1 | 6 | 676 | |||
| GenBank mRNA | AF368280.1 | - | 6 | 536 | ||
| FQ211527.1 | - | 6 | 579 | |||
| FQ212077.1 | - | 6 | 703 | |||
| FQ212215.1 | - | 6 | 678 | |||

Sgtb
Genomic Map