siRNA Details
| Gene Symbol: LOC100362324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002729007.4 | XP_002729053.2 | 23 | 3599 | ||
| XM_006231950.2 | XP_006232012.1 | 23 | 3599 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100362324| Gene Symbol: LOC100362324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002729007.4 | XP_002729053.2 | 23 | 3599 | ||
| XM_006231950.2 | XP_006232012.1 | 23 | 3599 | |||