siRNA Details
| Gene Symbol: Mocs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007633.2 | NP_001007634.1 | 5 | 637 | ||
| NM_001162413.1 | NP_001155885.1 | 6 | 759 | |||
| XM_006231985.2 | XP_006232047.1 | 6 | 705 | |||
| XM_006231986.2 | XP_006232048.1 | 5 | 602 | |||
| GenBank mRNA | BC079181.1 | - | 6 | 725 | 7130858 | |
| FM123325.1 | - | 5 | 591 | |||
| FQ220090.1 | - | 6 | 654 | |||
| FQ220210.1 | - | 5 | 550 | |||
| FQ230755.1 | - | 5 | 560 | |||
| FQ235152.1 | - | 6 | 688 | |||

Mocs2
Genomic Map