siRNA Details
| Gene Symbol: Pelo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007634.1 | NP_001007635.1 | 2 | 1086 | ||
| GenBank mRNA | BC083637.1 | - | 2 | 1086 | 7134766 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pelo| Gene Symbol: Pelo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007634.1 | NP_001007635.1 | 2 | 1086 | ||
| GenBank mRNA | BC083637.1 | - | 2 | 1086 | 7134766 | |