siRNA Details
| Gene Symbol: Mrps30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106412.1 | NP_001099882.1 | 2 | 716 | ||
| XM_006231957.2 | XP_006232019.1 | 2 | 736 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrps30| Gene Symbol: Mrps30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106412.1 | NP_001099882.1 | 2 | 716 | ||
| XM_006231957.2 | XP_006232019.1 | 2 | 736 | |||