siRNA Details
| Gene Symbol: Wdr70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013909.1 | NP_001013931.1 | 12 | 1246 | ||
| GenBank mRNA | BC089903.1 | - | 12 | 1246 | 7384104 | |
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 Wdr70
Wdr70| Gene Symbol: Wdr70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013909.1 | NP_001013931.1 | 12 | 1246 | ||
| GenBank mRNA | BC089903.1 | - | 12 | 1246 | 7384104 | |
 Back To Top
 Back To Top