siRNA Details
| Gene Symbol: Brix1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029915.1 | NP_001025086.1 | 9 | 747 | ||
| GenBank mRNA | BC098005.1 | - | 9 | 747 | 7453609 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Brix1| Gene Symbol: Brix1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029915.1 | NP_001025086.1 | 9 | 747 | ||
| GenBank mRNA | BC098005.1 | - | 9 | 747 | 7453609 | |