siRNA Details
| Gene Symbol: Rai14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011947.1 | NP_001011947.1 | 15, 16 | 2806 | ||
| XM_006232060.2 | XP_006232122.1 | 16, 17 | 3380 | |||
| XM_006232061.2 | XP_006232123.1 | 16, 17 | 2953 | |||
| XM_006232062.2 | XP_006232124.1 | 16, 17 | 3818 | |||
| XM_006232063.2 | XP_006232125.1 | 15, 16 | 3293 | |||
| XM_006232064.2 | XP_006232126.1 | 14, 15 | 3206 | |||
| XM_008760765.1 | XP_008758987.1 | 14, 15 | 3749 | |||
| GenBank mRNA | BC085775.1 | - | 15, 16 | 2806 | 7096978 | |

Rai14
Genomic Map