siRNA Details
| Gene Symbol: C1qtnf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006232067.2 | XP_006232129.2 | 1 | 65 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1qtnf3| Gene Symbol: C1qtnf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006232067.2 | XP_006232129.2 | 1 | 65 | ||