siRNA Details
| Gene Symbol: C1qtnf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134436.1 | NP_001127908.1 | 4, 5 | 767 | ||
| XM_006232067.2 | XP_006232129.2 | 4, 5 | 704 | |||
| GenBank mRNA | BC162048.1 | - | 4, 5 | 767 | 7108655 | |

C1qtnf3
Genomic Map