siRNA Details
| Gene Symbol: Tars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006976.1 | NP_001006977.1 | 17 | 1939 | ||
| XM_006232068.2 | XP_006232130.1 | 18 | 2097 | |||
| GenBank mRNA | BC083914.1 | - | 17 | 1939 | 7134406 | |
| FQ234975.1 | - | 5 | 407 | |||

Tars
Genomic Map