siRNA Details
| Gene Symbol: Tars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006976.1 | NP_001006977.1 | 10 | 1189 | ||
| XM_006232068.2 | XP_006232130.1 | 11 | 1347 | |||
| GenBank mRNA | BC083914.1 | - | 10 | 1189 | 7134406 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tars| Gene Symbol: Tars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006976.1 | NP_001006977.1 | 10 | 1189 | ||
| XM_006232068.2 | XP_006232130.1 | 11 | 1347 | |||
| GenBank mRNA | BC083914.1 | - | 10 | 1189 | 7134406 | |