siRNA Details
| Gene Symbol: Rgs9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019224.2 | NP_062097.1 | 12 | 957 | ||
| GenBank mRNA | AB019145.1 | - | 12 | 957 | ||
| AF038006.1 | - | 3 | 283 | |||
| AF071475.1 | - | 12 | 953 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rgs9| Gene Symbol: Rgs9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019224.2 | NP_062097.1 | 12 | 957 | ||
| GenBank mRNA | AB019145.1 | - | 12 | 957 | ||
| AF038006.1 | - | 3 | 283 | |||
| AF071475.1 | - | 12 | 953 | |||