siRNA Details
| Gene Symbol: Slc1a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289941.1 | NP_001276870.1 | 2 | 321 | ||
| NM_001289942.1 | NP_001276871.1 | 2 | 310 | |||
| NM_001289943.1 | NP_001276872.1 | 2 | 310 | |||
| NM_019225.2 | NP_062098.1 | 2 | 300 | |||
| GenBank mRNA | AF265360.1 | - | 2 | 160 | ||
| FQ097869.1 | - | 2 | 300 | |||
| S59158.1 | - | 2 | 242 | |||

Slc1a3
Genomic Map