siRNA Details
| Gene Symbol: Dnah5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760824.1 | XP_008759046.1 | 8031 | |||
| XM_226891.8 | XP_226891.6 | 12 | 2553 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnah5| Gene Symbol: Dnah5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760824.1 | XP_008759046.1 | 8031 | |||
| XM_226891.8 | XP_226891.6 | 12 | 2553 | |||