siRNA Details
| Gene Symbol: Slc7a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017217.1 | NP_058913.1 | 3 | 695 | ||
| XM_006257059.2 | XP_006257121.1 | 4 | 1025 | |||
| GenBank mRNA | AB000113.1 | - | 3 | 695 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc7a3| Gene Symbol: Slc7a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017217.1 | NP_058913.1 | 3 | 695 | ||
| XM_006257059.2 | XP_006257121.1 | 4 | 1025 | |||
| GenBank mRNA | AB000113.1 | - | 3 | 695 | ||