siRNA Details
| Gene Symbol: Cct5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004078.1 | NP_001004078.1 | 2, 3 | 179 | ||
| GenBank mRNA | AY683456.1 | - | 1, 2 | 61 | ||
| BC059165.1 | - | 2, 3 | 249 | |||
| BC079441.1 | - | 2, 3 | 179 | 7115414 | ||

Cct5
Genomic Map