siRNA Details
| Gene Symbol: Cct5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004078.1 | NP_001004078.1 | 7, 8 | 992 | ||
| GenBank mRNA | BC079441.1 | - | 7, 8 | 992 | 7115414 | |
| FQ229253.1 | - | 2, 3 | 211 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cct5| Gene Symbol: Cct5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004078.1 | NP_001004078.1 | 7, 8 | 992 | ||
| GenBank mRNA | BC079441.1 | - | 7, 8 | 992 | 7115414 | |
| FQ229253.1 | - | 2, 3 | 211 | |||