siRNA Details
| Gene Symbol: Cct5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004078.1 | NP_001004078.1 | 4 | 467 | ||
| GenBank mRNA | BC059165.1 | - | 4 | 537 | ||
| BC079441.1 | - | 4 | 467 | 7115414 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cct5| Gene Symbol: Cct5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004078.1 | NP_001004078.1 | 4 | 467 | ||
| GenBank mRNA | BC059165.1 | - | 4 | 537 | ||
| BC079441.1 | - | 4 | 467 | 7115414 | ||