siRNA Details
| Gene Symbol: Gstt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012796.2 | NP_036928.1 | 2 | 273 | ||
| GenBank mRNA | BC061856.1 | - | 2 | 277 | 5623709 | |
| CK473092.1 | - | 2 | 273 | |||
| D10026.1 | - | 2 | 261 | |||
| FJ179409.1 | - | 2 | 139 | |||
| Gene Symbol: LOC103694878 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773847.1 | XP_008772069.1 | 2 | 493 | ||

Gstt2
Genomic Map