siRNA Details
| Gene Symbol: Gstt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012796.2 | NP_036928.1 | 3 | 346 | ||
| GenBank mRNA | BC061856.1 | - | 3 | 350 | 5623709 | |
| CK473092.1 | - | 3 | 346 | |||
| D10026.1 | - | 3 | 334 | |||
| FJ179409.1 | - | 3 | 212 | |||
| Gene Symbol: LOC103694879 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773848.1 | XP_008772070.1 | 1 | 416 | ||

Gstt2
Genomic Map