siRNA Details
Gene Symbol: LOC103694878 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773847.1 | XP_008772069.1 | 2 | 539 |
Gene Symbol: Gstt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012796.2 | NP_036928.1 | 2 | 319 | ||
GenBank mRNA | BC061856.1 | - | 2 | 323 | 5623709 | |
CK473092.1 | - | 2 | 319 | |||
D10026.1 | - | 2 | 307 | |||
FJ179409.1 | - | 2 | 185 |
