siRNA Details
| Gene Symbol: Dync1h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019226.3 | NP_062099.3 | 75 | 13544 | ||
| GenBank mRNA | L08505.1 | - | 75 | 13544 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dync1h1| Gene Symbol: Dync1h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019226.3 | NP_062099.3 | 75 | 13544 | ||
| GenBank mRNA | L08505.1 | - | 75 | 13544 | ||