siRNA Details
| Gene Symbol: Sec62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034129.1 | NP_001029301.1 | 6 | 781 | ||
| XM_006232224.2 | XP_006232286.1 | 2 | 363 | |||
| GenBank mRNA | AY325207.1 | - | 6 | 781 | ||
| FQ220436.1 | - | 2 | 140 | |||

Sec62
Genomic Map