siRNA Details
| Gene Symbol: Sec62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034129.1 | NP_001029301.1 | 3 | 245 | ||
| GenBank mRNA | AY325207.1 | - | 3 | 245 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sec62| Gene Symbol: Sec62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034129.1 | NP_001029301.1 | 3 | 245 | ||
| GenBank mRNA | AY325207.1 | - | 3 | 245 | ||