siRNA Details
| Gene Symbol: Mecom | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106423.1 | NP_001099893.1 | 13 | 3624 | ||
| XM_006232249.2 | XP_006232311.1 | 13 | 3432 | |||
| XM_006232250.2 | XP_006232312.1 | 13 | 3674 | |||
| XM_008760907.1 | XP_008759129.1 | 13 | 3429 | |||
| XM_008760908.1 | XP_008759130.1 | 12 | 3405 | |||
| XM_008760909.1 | XP_008759131.1 | 12 | 3401 | |||
| XM_008760910.1 | XP_008759132.1 | 13 | 3431 | |||
| XM_008760911.1 | XP_008759133.1 | 12 | 2429 | |||

Mecom
Genomic Map