siRNA Details
| Gene Symbol: Kcnmb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176861.1 | NP_789831.1 | 4 | 479 | ||
| GenBank mRNA | AB193523.1 | - | 2 | 112 | ||
| AJ517198.1 | - | 3 | 363 | |||
| AY191836.1 | - | 4 | 479 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnmb2| Gene Symbol: Kcnmb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176861.1 | NP_789831.1 | 4 | 479 | ||
| GenBank mRNA | AB193523.1 | - | 2 | 112 | ||
| AJ517198.1 | - | 3 | 363 | |||
| AY191836.1 | - | 4 | 479 | |||