siRNA Details
| Gene Symbol: Ndufb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106426.1 | NP_001099896.1 | 2 | 245 | ||
| GenBank mRNA | FQ224246.1 | - | 2 | 196 | ||
| FQ224437.1 | - | 2 | 198 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ndufb5| Gene Symbol: Ndufb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106426.1 | NP_001099896.1 | 2 | 245 | ||
| GenBank mRNA | FQ224246.1 | - | 2 | 196 | ||
| FQ224437.1 | - | 2 | 198 | |||