siRNA Details
Gene Symbol: Ndufb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106426.1 | NP_001099896.1 | 5 | 436 | ||
GenBank mRNA | FQ224246.1 | - | 5 | 387 | ||
FQ224437.1 | - | 5 | 389 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ndufb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106426.1 | NP_001099896.1 | 5 | 436 | ||
GenBank mRNA | FQ224246.1 | - | 5 | 387 | ||
FQ224437.1 | - | 5 | 389 |