siRNA Details
| Gene Symbol: Mccc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009653.1 | NP_001009653.1 | 11 | 1244 | ||
| GenBank mRNA | BC088473.1 | - | 11 | 1244 | 7128872 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mccc1| Gene Symbol: Mccc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009653.1 | NP_001009653.1 | 11 | 1244 | ||
| GenBank mRNA | BC088473.1 | - | 11 | 1244 | 7128872 | |