siRNA Details
| Gene Symbol: Exosc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025406.1 | NP_001020577.1 | 10 | 1064 | ||
| XM_008760913.1 | XP_008759135.1 | 10 | 1108 | |||
| GenBank mRNA | BC097413.1 | - | 10 | 1064 | 7454278 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Exosc9| Gene Symbol: Exosc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025406.1 | NP_001020577.1 | 10 | 1064 | ||
| XM_008760913.1 | XP_008759135.1 | 10 | 1108 | |||
| GenBank mRNA | BC097413.1 | - | 10 | 1064 | 7454278 | |